siRNA Details
| Gene Symbol: Cadm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145977.1 | NP_001139449.1 | 4 | 1103 | ||
| NM_178721.4 | NP_848836.2 | 5 | 1130 | |||
| GenBank mRNA | AK033973.1 | - | 4 | 730 | ||
| AK038842.1 | - | 5 | 1109 | |||
| AK043760.1 | - | 5 | 1113 | |||
| AK046800.1 | - | 4 | 1006 | |||
| AK163636.1 | - | 5 | 1107 | |||
| AK165030.1 | - | 5 | 1127 | |||

Cadm2
Genomic Map