siRNA Details
| Gene Symbol: Ptpn21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146199.1 | NP_001139671.1 | 12 | 1261 | ||
| NM_011877.2 | NP_036007.2 | 12 | 1396 | |||
| GenBank mRNA | BC014691.1 | - | 12 | 1526 | ||
| BC066856.1 | - | 11 | 1199 | |||
| BC098500.1 | - | 12 | 1261 | 30081068 | ||
| D37801.1 | - | 12 | 1345 | |||

Ptpn21
Genomic Map