siRNA Details
| Gene Symbol: Ptpn21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146199.1 | NP_001139671.1 | 2 | 236 | ||
| NM_011877.2 | NP_036007.2 | 2 | 371 | |||
| GenBank mRNA | AK013777.1 | - | 2 | 373 | ||
| AK030456.1 | - | 2 | 372 | |||
| BC014691.1 | - | 2 | 501 | |||
| BC049893.1 | - | 1 | 203 | |||
| BC066856.1 | - | 1 | 174 | |||
| D37801.1 | - | 2 | 320 | |||

Ptpn21
Genomic Map