siRNA Details
| Gene Symbol: Ptpn21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049893.1 | - | 7 | 897 | ||
| D37801.1 | - | 8 | 1014 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn21| Gene Symbol: Ptpn21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049893.1 | - | 7 | 897 | ||
| D37801.1 | - | 8 | 1014 | |||