siRNA Details
| Gene Symbol: Tiam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122998.1 | NP_001116470.1 | 22 | 4433 | ||
| NM_001286757.1 | NP_001273686.1 | 8 | 1476 | |||
| NM_001286758.1 | NP_001273687.1 | 8 | 840 | |||
| NM_011878.2 | NP_036008.2 | 22 | 4417 | |||
| GenBank mRNA | AB022915.1 | - | 22 | 4418 | ||
| AF121102.1 | - | 3 | 415 | |||
| AK129474.1 | - | 22 | 4336 | |||
| AK158489.1 | - | 8 | 841 | |||
| AK161947.1 | - | 8 | 1477 | |||
| AK162644.1 | - | 8 | 1153 | |||
| BC067048.1 | - | 22 | 4430 | 30535152 | ||
| BC079600.1 | - | 22 | 4322 | 6853027 | ||

Tiam2
Genomic Map