siRNA Details
| Gene Symbol: Tiam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122998.1 | NP_001116470.1 | 23 | 4467 | ||
| NM_001286757.1 | NP_001273686.1 | 9 | 1510 | |||
| NM_001286758.1 | NP_001273687.1 | 9 | 874 | |||
| NM_011878.2 | NP_036008.2 | 23 | 4451 | |||
| GenBank mRNA | AB022915.1 | - | 23 | 4452 | ||
| AF121102.1 | - | 4 | 449 | |||
| AK129474.1 | - | 23 | 4370 | |||
| AK158489.1 | - | 9 | 875 | |||
| AK161947.1 | - | 9 | 1511 | |||
| AK162644.1 | - | 9 | 1187 | |||
| BC067048.1 | - | 23 | 4464 | 30535152 | ||
| BC079600.1 | - | 23 | 4356 | 6853027 | ||

Tiam2
Genomic Map