siRNA Details
| Gene Symbol: Zfp870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207245.2 | NP_997128.1 | 4 | 802 | ||
| GenBank mRNA | BC066107.1 | - | 4 | 726 | 30537752 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp870| Gene Symbol: Zfp870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207245.2 | NP_997128.1 | 4 | 802 | ||
| GenBank mRNA | BC066107.1 | - | 4 | 726 | 30537752 | |