siRNA Details
| Gene Symbol: Btnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111094.1 | NP_001104564.1 | 4 | 827 | ||
| GenBank mRNA | BC052925.1 | - | 4 | 805 | 6398253 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btnl1| Gene Symbol: Btnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111094.1 | NP_001104564.1 | 4 | 827 | ||
| GenBank mRNA | BC052925.1 | - | 4 | 805 | 6398253 | |