siRNA Details
| Gene Symbol: Cchcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146248.2 | NP_666360.2 | 4 | 627 | ||
| GenBank mRNA | AK159780.1 | - | 4 | 554 | ||
| BC031416.1 | - | 5 | 716 | 4012768 | ||
| BM947393.1 | - | 4 | 539 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cchcr1| Gene Symbol: Cchcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146248.2 | NP_666360.2 | 4 | 627 | ||
| GenBank mRNA | AK159780.1 | - | 4 | 554 | ||
| BC031416.1 | - | 5 | 716 | 4012768 | ||
| BM947393.1 | - | 4 | 539 | |||