siRNA Details
| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010833.2 | NP_001010833.2 | 10 | 4149 | ||
| GenBank mRNA | AK129074.1 | - | 4 | 1885 | ||
| BC085140.1 | - | 5 | 2018 | |||
| BC094363.1 | - | 6 | 2852 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mdc1| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010833.2 | NP_001010833.2 | 10 | 4149 | ||
| GenBank mRNA | AK129074.1 | - | 4 | 1885 | ||
| BC085140.1 | - | 5 | 2018 | |||
| BC094363.1 | - | 6 | 2852 | |||