siRNA Details
| Gene Symbol: Rasgrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166493.1 | NP_001159965.1 | 13 | 1873 | ||
| NM_207246.4 | NP_997129.1 | 14 | 1865 | |||
| GenBank mRNA | AB154824.1 | - | 12 | 1560 | ||
| AK173041.1 | - | 13 | 1758 | |||
| BC066069.1 | - | 12 | 1503 | 6851971 | ||
| BC066127.1 | - | 14 | 1790 | 30280332 | ||

Rasgrp3
Genomic Map