siRNA Details
| Gene Symbol: Rnf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF037205.1 | - | 7 | 601 | ||
| AF037206.1 | - | 7 | 601 | |||
| AK034135.1 | - | 7 | 662 | |||
| AK036591.1 | - | 7 | 646 | |||
| AK050095.1 | - | 7 | 1346 | |||
| AK082296.1 | - | 7 | 642 | |||
| AK139447.1 | - | 7 | 607 | |||
| AK140682.1 | - | 7 | 656 | |||
| AK158046.1 | - | 7 | 642 | |||
| AK167278.1 | - | 7 | 606 | |||
| BC058182.1 | - | 7 | 609 | 5324040 | ||

Rnf13
Genomic Map