siRNA Details
| Gene Symbol: 9430020K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081963.1 | NP_001075432.1 | 3 | 2333 | ||
| GenBank mRNA | AK220394.1 | - | 1 | 490 | ||
| BC144919.1 | - | 2 | 2237 | 9053467 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9430020K01Rik| Gene Symbol: 9430020K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081963.1 | NP_001075432.1 | 3 | 2333 | ||
| GenBank mRNA | AK220394.1 | - | 1 | 490 | ||
| BC144919.1 | - | 2 | 2237 | 9053467 | ||