siRNA Details
| Gene Symbol: Zfp438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178722.5 | NP_848837.3 | 4 | 300 | ||
| GenBank mRNA | AK034745.1 | - | 4 | 301 | ||
| AK046812.1 | - | 4 | 296 | |||
| AK084655.1 | - | 4 | 297 | |||
| BC096460.1 | - | 4 | 300 | 5374843 | ||

Zfp438
Genomic Map