siRNA Details
| Gene Symbol: E230025N22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172831.2 | NP_766419.2 | 7 | 699 | ||
| GenBank mRNA | AK087618.1 | - | 7 | 703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E230025N22Rik| Gene Symbol: E230025N22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172831.2 | NP_766419.2 | 7 | 699 | ||
| GenBank mRNA | AK087618.1 | - | 7 | 703 | ||