siRNA Details
| Gene Symbol: Fem1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173423.4 | NP_775599.1 | 3 | 1361 | ||
| GenBank mRNA | AK028330.1 | - | 3 | 1362 | ||
| AK049336.1 | - | 3 | 1349 | |||
| AK129440.1 | - | 1294 | ||||
| AK147729.1 | - | 3 | 1251 | |||
| AY249189.1 | - | 2 | 823 | |||
| BC138604.1 | - | 2 | 904 | 8861624 | ||
| BC138605.1 | - | 2 | 904 | 8861625 | ||

Fem1c
Genomic Map