siRNA Details
| Gene Symbol: Adamts19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175506.3 | NP_780715.1 | 21 | 3300 | ||
| GenBank mRNA | AK029178.1 | - | 11 | 2767 | ||
| AY135183.1 | - | 21 | 3300 | |||
| BC150736.1 | - | 21 | 3397 | 9087647 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts19| Gene Symbol: Adamts19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175506.3 | NP_780715.1 | 21 | 3300 | ||
| GenBank mRNA | AK029178.1 | - | 11 | 2767 | ||
| AY135183.1 | - | 21 | 3300 | |||
| BC150736.1 | - | 21 | 3397 | 9087647 | ||