siRNA Details
| Gene Symbol: Gm4951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033767.3 | NP_001028939.1 | 3 | 609 | ||
| GenBank mRNA | AK149556.1 | - | 2 | 501 | ||
| AK149583.1 | - | 3 | 607 | |||
| BC139145.1 | - | 2 | 437 | 8862167 | ||
| BC139147.1 | - | 2 | 437 | 8862169 | ||

Gm4951
Genomic Map