siRNA Details
| Gene Symbol: Sgcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011891.4 | NP_036021.1 | 6 | 580 | ||
| GenBank mRNA | AB024923.1 | - | 6 | 801 | ||
| AK040974.1 | - | 6 | 582 | |||
| AK047384.1 | - | 1322 | ||||
| BC130019.1 | - | 6 | 570 | 40140624 | ||

Sgcd
Genomic Map