siRNA Details
| Gene Symbol: Sgcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011891.4 | NP_036021.1 | 3 | 294 | ||
| GenBank mRNA | AB024923.1 | - | 3 | 515 | ||
| AK040974.1 | - | 3 | 296 | |||
| BC130019.1 | - | 3 | 284 | 40140624 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgcd| Gene Symbol: Sgcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011891.4 | NP_036021.1 | 3 | 294 | ||
| GenBank mRNA | AB024923.1 | - | 3 | 515 | ||
| AK040974.1 | - | 3 | 296 | |||
| BC130019.1 | - | 3 | 284 | 40140624 | ||