siRNA Details
| Gene Symbol: Sh3bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155103.1 | - | 9 | 1394 | ||
| AK155970.1 | - | 9 | 1485 | |||
| BC010198.1 | - | 9 | 1375 | 3967543 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3bp2| Gene Symbol: Sh3bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155103.1 | - | 9 | 1394 | ||
| AK155970.1 | - | 9 | 1485 | |||
| BC010198.1 | - | 9 | 1375 | 3967543 | ||