siRNA Details
| Gene Symbol: Smc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019710.2 | NP_062684.2 | 11 | 1884 | ||
| GenBank mRNA | AF047600.1 | - | 11 | 1884 | ||
| AK145255.1 | - | 9 | 1516 | |||
| BC003279.1 | - | 11 | 1821 | |||
| BC025590.1 | - | 11 | 1827 | |||
| BC036973.1 | - | 11 | 1837 | |||
| BC131667.1 | - | 11 | 1837 | 40110822 | ||

Smc1a
Genomic Map