siRNA Details
| Gene Symbol: Smc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019710.2 | NP_062684.2 | 9 | 1570 | ||
| GenBank mRNA | AF047600.1 | - | 9 | 1570 | ||
| AK145255.1 | - | 7 | 1202 | |||
| BC003279.1 | - | 9 | 1507 | |||
| BC025590.1 | - | 9 | 1513 | |||
| BC036973.1 | - | 9 | 1523 | |||
| BC131667.1 | - | 9 | 1523 | 40110822 | ||

Smc1a
Genomic Map