siRNA Details
Gene Symbol: Sec31b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033343.1 | NP_001028515.1 | 3 | 192 | ||
GenBank mRNA | BB626493.1 | - | 3 | 193 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sec31b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033343.1 | NP_001028515.1 | 3 | 192 | ||
GenBank mRNA | BB626493.1 | - | 3 | 193 |