siRNA Details
| Gene Symbol: Sec31b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033343.1 | NP_001028515.1 | 11 | 1309 | ||
| GenBank mRNA | AK158017.1 | - | 10 | 1186 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec31b| Gene Symbol: Sec31b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033343.1 | NP_001028515.1 | 11 | 1309 | ||
| GenBank mRNA | AK158017.1 | - | 10 | 1186 | ||