siRNA Details
| Gene Symbol: Sufu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025391.2 | NP_001020562.1 | 4, 5 | 795 | ||
| NM_015752.3 | NP_056567.2 | 4, 5 | 795 | |||
| GenBank mRNA | AF134893.1 | - | 4, 5 | 735 | ||
| AJ131692.1 | - | 4, 5 | 796 | |||
| AJ308625.1 | - | 4, 5 | 474 | |||
| AJ308626.1 | - | 4, 5 | 729 | |||
| AK015885.1 | - | 3, 4 | 553 | |||
| AK016976.1 | - | 4, 5 | 774 | |||
| AK047603.1 | - | 4, 5 | 665 | |||
| AK156410.1 | - | 4, 5 | 757 | |||
| BC048168.1 | - | 4, 5 | 768 | 6821477 | ||
| BC056997.1 | - | 4, 5 | 768 | 6821477 | ||

Sufu
Genomic Map