siRNA Details
| Gene Symbol: Tor1aip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172843.4 | NP_766431.3 | 6 | 1660 | ||
| GenBank mRNA | AK038024.1 | - | 6 | 1922 | ||
| AK080959.1 | - | 4 | 939 | |||
| BC066790.1 | - | 4 | 866 | 30099444 | ||
| CK635751.1 | - | 2 | 501 | |||

Tor1aip2
Genomic Map