siRNA Details
| Gene Symbol: Tlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112705.2 | NP_001106176.1 | 9, 10 | 1256 | ||
| NM_001294329.1 | NP_001281258.1 | 7, 8 | 1116 | |||
| NM_001294330.1 | NP_001281259.1 | 7, 8 | 1064 | |||
| NM_001294331.1 | NP_001281260.1 | 9, 10 | 1269 | |||
| NM_001294334.1 | NP_001281263.1 | 9, 10 | 1256 | |||
| GenBank mRNA | AF045252.1 | - | 9, 10 | 1275 | ||
| AF045253.1 | - | 8, 9 | 703 | |||
| AF045254.1 | - | 4, 5 | 417 | |||
| AK014829.1 | - | 9, 10 | 894 | |||
| AK089925.1 | - | 7, 8 | 644 | |||
| AK165667.1 | - | 9, 10 | 1259 | |||
| AX775743.1 | - | 3, 4 | 281 | |||
| BC066198.1 | - | 7, 8 | 609 | |||
| CD554252.1 | - | 3, 4 | 380 | |||

Tlk2
Genomic Map