siRNA Details
| Gene Symbol: Tll2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011904.3 | NP_036034.1 | 2 | 344 | ||
| GenBank mRNA | AF073526.1 | - | 2 | 405 | ||
| BC132537.1 | - | 2 | 344 | 40130814 | ||
| BC145132.1 | - | 2 | 344 | 9053680 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tll2| Gene Symbol: Tll2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011904.3 | NP_036034.1 | 2 | 344 | ||
| GenBank mRNA | AF073526.1 | - | 2 | 405 | ||
| BC132537.1 | - | 2 | 344 | 40130814 | ||
| BC145132.1 | - | 2 | 344 | 9053680 | ||