siRNA Details
Gene Symbol: Fmo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172844.2 | NP_766432.1 | 3 | 225 | ||
GenBank mRNA | AK029219.1 | - | 3 | 226 | ||
AK161224.1 | - | 3 | 225 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fmo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172844.2 | NP_766432.1 | 3 | 225 | ||
GenBank mRNA | AK029219.1 | - | 3 | 226 | ||
AK161224.1 | - | 3 | 225 |