siRNA Details
| Gene Symbol: Tnfsf13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033622.1 | NP_296371.1 | 6 | 934 | ||
| GenBank mRNA | AF119383.1 | - | 6 | 934 | ||
| AF352245.1 | - | 6 | 938 | |||
| AK034121.1 | - | 6 | 566 | |||
| AK079180.1 | - | 6 | 1153 | |||
| AK155288.1 | - | 6 | 713 | |||
| AY290823.1 | - | 5 | 661 | |||
| BC106840.1 | - | 5 | 850 | 40045718 | ||
| BC106841.1 | - | 6 | 907 | 40045720 | ||

Tnfsf13b
Genomic Map