siRNA Details
| Gene Symbol: Abca12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175210.3 | NP_780419.2 | 21 | 3064 | ||
| GenBank mRNA | AK076440.1 | - | 2 | 115 | ||
| BC158053.1 | - | 21 | 2877 | 9088162 | ||
| BC158063.1 | - | 21 | 2877 | 9007362 | ||

Abca12
Genomic Map