siRNA Details
| Gene Symbol: Catip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033345.2 | NP_001028517.1 | 2 | 93 | ||
| GenBank mRNA | AK138157.1 | - | 2 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Catip| Gene Symbol: Catip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033345.2 | NP_001028517.1 | 2 | 93 | ||
| GenBank mRNA | AK138157.1 | - | 2 | 94 | ||