siRNA Details
Gene Symbol: Nyap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172849.3 | NP_766437.2 | 5 | 2749 | ||
GenBank mRNA | BC082310.1 | - | 5 | 2749 | 30355797 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nyap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172849.3 | NP_766437.2 | 5 | 2749 | ||
GenBank mRNA | BC082310.1 | - | 5 | 2749 | 30355797 |