siRNA Details
| Gene Symbol: Nyap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172849.3 | NP_766437.2 | 5 | 2702 | ||
| GenBank mRNA | BC082310.1 | - | 5 | 2702 | 30355797 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nyap2| Gene Symbol: Nyap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172849.3 | NP_766437.2 | 5 | 2702 | ||
| GenBank mRNA | BC082310.1 | - | 5 | 2702 | 30355797 | |