siRNA Details
| Gene Symbol: Ankmy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172850.3 | NP_766438.2 | 16, 17 | 2880 | ||
| GenBank mRNA | AK029700.1 | - | 16, 17 | 2885 | ||
| BC120887.1 | - | 15, 16 | 2716 | 40060916 | ||
| BC120888.1 | - | 15, 16 | 2719 | 40060917 | ||

Ankmy1
Genomic Map