siRNA Details
| Gene Symbol: Cntnap5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172851.2 | NP_766439.2 | 13 | 2052 | ||
| GenBank mRNA | AK082656.1 | - | 7 | 1110 | ||
| AK133394.1 | - | 3 | 382 | |||
| BC145618.1 | - | 6 | 817 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntnap5b| Gene Symbol: Cntnap5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172851.2 | NP_766439.2 | 13 | 2052 | ||
| GenBank mRNA | AK082656.1 | - | 7 | 1110 | ||
| AK133394.1 | - | 3 | 382 | |||
| BC145618.1 | - | 6 | 817 | |||