siRNA Details
Gene Symbol: Itga8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001309.2 | NP_001001309.1 | 2 | 385 | ||
GenBank mRNA | AF041409.1 | - | 2 | 116 | ||
AK044910.1 | - | 2 | 387 | |||
AK135193.1 | - | 2 | 364 | |||
BC137793.1 | - | 2 | 357 |
