siRNA Details
| Gene Symbol: Lrrc8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177725.4 | NP_808393.1 | 4 | 2366 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc8a| Gene Symbol: Lrrc8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177725.4 | NP_808393.1 | 4 | 2366 | ||