siRNA Details
| Gene Symbol: Zeb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289521.1 | NP_001276450.1 | 9 | 3273 | ||
| NM_015753.4 | NP_056568.2 | 10 | 3610 | |||
| GenBank mRNA | AF033116.1 | - | 9 | 3125 | ||
| AK032970.1 | - | 3 | 411 | |||
| AK122312.1 | - | 10 | 3183 | |||
| AK141157.1 | - | 9 | 3241 | |||
| BC060699.1 | - | 10 | 3347 | 6408845 | ||

Zeb2
Genomic Map