siRNA Details
| Gene Symbol: D430041D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033347.2 | NP_001028519.2 | 5 | 1589 | ||
| GenBank mRNA | AI606651.1 | - | 1 | 94 | ||
| BX517992.1 | - | 1 | 95 | |||
| CX239849.1 | - | 2 | 165 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D430041D05Rik| Gene Symbol: D430041D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033347.2 | NP_001028519.2 | 5 | 1589 | ||
| GenBank mRNA | AI606651.1 | - | 1 | 94 | ||
| BX517992.1 | - | 1 | 95 | |||
| CX239849.1 | - | 2 | 165 | |||