siRNA Details
| Gene Symbol: Fermt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198029.2 | NP_932146.2 | 13 | 2385 | ||
| GenBank mRNA | AF073489.1 | - | 10 | 1171 | ||
| AK030947.1 | - | 12 | 2575 | |||
| AK050804.1 | - | 12 | 1533 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fermt1| Gene Symbol: Fermt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198029.2 | NP_932146.2 | 13 | 2385 | ||
| GenBank mRNA | AF073489.1 | - | 10 | 1171 | ||
| AK030947.1 | - | 12 | 2575 | |||
| AK050804.1 | - | 12 | 1533 | |||