siRNA Details
| Gene Symbol: Dzank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172859.2 | NP_766447.2 | 8 | 839 | ||
| GenBank mRNA | AK032182.1 | - | 8 | 839 | ||
| AK032632.1 | - | 8 | 830 | |||
| AK163994.1 | - | 8 | 830 | |||
| BC141156.1 | - | 8 | 738 | 9055768 | ||
| BC145464.1 | - | 8 | 738 | 9054026 | ||

Dzank1
Genomic Map