siRNA Details
| Gene Symbol: L3mbtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081338.1 | NP_001074807.1 | 14 | 1683 | ||
| GenBank mRNA | AK049859.1 | - | 14 | 1738 | ||
| AK220350.1 | - | 12 | 1431 | |||
| BC116639.1 | - | 13 | 1642 | 40061282 | ||

L3mbtl1
Genomic Map