siRNA Details
| Gene Symbol: Ankub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033349.2 | NP_001028521.1 | 2 | 300 | ||
| GenBank mRNA | AK134766.1 | - | 2 | 257 | ||
| AK138483.1 | - | 2 | 300 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankub1| Gene Symbol: Ankub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033349.2 | NP_001028521.1 | 2 | 300 | ||
| GenBank mRNA | AK134766.1 | - | 2 | 257 | ||
| AK138483.1 | - | 2 | 300 | |||