siRNA Details
| Gene Symbol: Rxfp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181817.1 | NP_861538.1 | 1 | 227 | ||
| GenBank mRNA | AY255539.1 | - | 1 | 107 | ||
| AY288422.1 | - | 1 | 227 | |||
| AY633765.1 | - | 1 | 227 | |||
| BC107269.1 | - | 1 | 227 | 40052601 | ||
| BC107270.1 | - | 1 | 226 | |||
More Information
Set Membership: |
|

Rxfp4
Genomic Map