siRNA Details
| Gene Symbol: Bank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033350.2 | NP_001028522.2 | 9 | 1673 | ||
| GenBank mRNA | AA177599.1 | - | 1 | 208 | ||
| AK156201.1 | - | 9 | 1683 | |||
| AY178734.1 | - | 9 | 1693 | |||
| BC026932.1 | - | 9 | 1620 | 4008261 | ||
| EU051378.1 | - | 8 | 1274 | |||

Bank1
Genomic Map