siRNA Details
| Gene Symbol: Bank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033350.2 | NP_001028522.2 | 12 | 2226 | ||
| GenBank mRNA | AK041039.1 | - | 3 | 244 | ||
| AK148665.1 | - | 1 | 21 | |||
| AK156201.1 | - | 12 | 2236 | |||
| AY178734.1 | - | 12 | 2246 | |||
| BC026932.1 | - | 12 | 2173 | 4008261 | ||
| BC116328.1 | - | 2 | 309 | |||
| BE848854.1 | - | 3 | 380 | |||
| EU051378.1 | - | 11 | 1827 | |||

Bank1
Genomic Map