siRNA Details
| Gene Symbol: Manea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172865.2 | NP_766453.2 | 5 | 1448 | ||
| GenBank mRNA | AK030141.1 | - | 5 | 1423 | ||
| BC067076.1 | - | 5 | 1448 | 30628555 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Manea| Gene Symbol: Manea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172865.2 | NP_766453.2 | 5 | 1448 | ||
| GenBank mRNA | AK030141.1 | - | 5 | 1423 | ||
| BC067076.1 | - | 5 | 1448 | 30628555 | ||