siRNA Details
| Gene Symbol: Frmpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081172.2 | NP_001074641.1 | 15 | 1647 | ||
| GenBank mRNA | AK122409.1 | - | 12 | 1273 | ||
| AK133351.1 | - | 2 | 86 | |||
| BC031840.1 | - | 5 | 507 | |||
| BC051097.1 | - | 5 | 507 | |||
| BC158060.1 | - | 15 | 1611 | 9007360 | ||

Frmpd1
Genomic Map